missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Life Sciences Protein G GraviTrap™

Includes ten ready-to-use columns prepacked with 1ml of Protein G Sepharose 4 Fast Flow

Brand:  Cytiva 28-9852-55

870.13 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.



Product Code. 12526518

  • 980.00 € / 10mL
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Prepacked gravity-flow columns provide convenience, save time, and increase the reliability of your results.

  • Purification of antibodies in 30 min allows you to quickly obtain and store your target protein in order to avoid the inherent dangers of protein instability and degradation associated with slow and long purification runs
  • High purity and recovery with purity > 95% in one step and recovery typically 70% to 80%
  • High binding capacity such as Protein G GraviTrap approximately 20mg human IgG/column

  • Complete pack size: 10 columns
  • Application: Manual purification of monoclonal and polyclonal antibodies
  • Includes: 1 pack of 10 × 1mL columns
  • Binding capacity: Approx. 20mg human IgG/column
  • Bed volume: 1mL
  • Material (Column hardware): Polypropylene barrel, Polyethylene frits

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 mL
Protein G GraviTrap includes ten columns with 1 mL of resin for general purpose purification of polyclonal and monoclonal antibodies from most species.
2°C to 8°C
Fast and efficient manual purification of monoclonal and polyclonal antibodies
1 μL
10 Ready-to -use column prepacked with 1mL of Protein G Sepharose 4 fast flow, Polypropylene barrel, Polyethylene frits
10 Pack
Gravity Flow Column
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • protein G gravitrap

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235-Keep cool.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers