Learn More
Invitrogen™ Coverglass Removal Tool
Minimize the risk of breakage of chambered coverglass during cell culture and fluorescence imaging applications. CultureWell&Trade; Coverglass Removal Tool enables the chambered upper structure to be easily separated from the coverglass without the need for excessive force.
Brand: Invitrogen™ C37010
20.41 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
The Coverglass Removal Tool is intended for use with a CultureWell™ Chambered Coverglass for Cell Culture (Mfc. Nos. C37000, C37005). The removable chambered coverglass is provided sterile and ready to use. It contains 16 wells that can each hold up to 250μL, allowing cells to be cultured in a number of different conditions on a single slide. When the cells are ready to be imaged, coverglass removal is made easy by the use of this simple tool.
Specifications
Coverglass Removal Tool | |
Culturewell | |
Microscope | |
1 each |