Learn More
Invitrogen™ E-Gel™ Imager Quantitation USB Dongle
Key to operating Gel Quant Express software on one or more computers
Brand:  Invitrogen™ 4466610
Product Code. 11642129
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Includes: 1 x Gel Quant Express software enabling USB Dongle
Description
Description
To use this powerful quantitation and analysis software, insert an activation dongle into a free USB port on PC whenever the software is in use. With additional USB dongles, software can be used on more than one computer simultaneously.
  • Access powerful Gel Quant Express software using USB dongle
  • Key to Quantitation Gel Quant Express is powerful and intuitive software tool included with each E-Gel Imager system
  • In order to utilize software, HASP key in form of USB dongle must be inserted into free USB port on computer
  • Each E-Gel Imager system includes single Quantitation Software USB Dongle that will allow single instance of Gel quant Express software to be used at given time
  • Separate USB dongle is required for each instance of Gel Quant Express Software that will be used at given time
  • Compact Solution for Agarose Gel Imaging, Analysis, and Documentation
  • Powerful and flexible system that allows to document gel images using multiple nucleic acid dyes and even Qdottrademark (TM) nanocrystals
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 x Gel Quant Express software enabling USB Dongle.
Store at Room temperature.
E-Gel
Windows XP (Service Pack 3), Windows 7
Validation and Verification Software
E-Gel Imager
Imager Quantitation USB Dongle
Room Temperature
Image Analysis
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.