missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBind™ Fluorescent Detection (FD) Solution Kit

Optimized for western blotting with fluorescent-conjugated antibodies. Invitrogen™ iBind™ Fluorescent Detection (FD) Solution Kit is intended for use with the iBind™ Flex Western Device (SLF2000) for use in infrared detection and dual wavelength semi-quantitative analysis.

Brand:  Invitrogen™ SLF1019

67.64 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 15297029

  • 89.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Each iBind™ Fluorescent Detection (FD) Solution Kit contains sufficient solution for 10 mini-membrane reactions. An iBind™ Western Device and iBind™ Cards are required for use of this kit.

  • Optimized solutions—Ideal viscosity for blocking, washing and antibody dilution
  • Long shelf life—Store iBind™ FD Solution Kit for 12 months at 4°C
  • Better antigen-antibody interaction—Solutions flow between the iBind™ Card and membrane for maximum interaction between protein and antibody

About the iBind™ Western Device:
The iBind™ Western Device is an automated western-processing device that performs every step from blocking to washing to antibody incubations via sequential lateral flow (SLF). SLF allows the timely release and flow of solutions and antibodies to the membrane without need of an external power source. Each solution is released from the iBind™ Device wells to an iBind™ Card via SLF. The solutions are then wicked toward the stack region of the card. The glass fiber matrix of the card allows for homogenous and consistent flow of the solutions to the membrane, increasing the antigen-antibody interaction.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Fluroescent Detection Kit
Western Blot
iBind™
10 mini-membrane reactions
Contains: 1 bottle 5X buffer, 2 vials 100X additive, 1 vial 10% SDS

Store at 2–8°C.
Fluorescence
Nitrocellulose, PVDF
1 kit
1 bottle 5X buffer, 2 vials 100X additive, 1 vial 10% SDS
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.