Learn More
Applied Biosystems™ TaqMan™ RNase P Instrument Verification Plate, 96-well (for 0.2 mL block)
Used to validate the performance of the Applied Biosystems™ ViiA™ 7 Real-Time PCR Systems with the 96-well Block.
Brand: Applied Biosystems™ 4432382
478.80 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Description
The TaqMan RNase P Instrument Verification Kit is used to validate the performance of the Applied Biosystems real-time PCR systems with the 96-well block.
The plate preloaded with the necessary reagents to detect and quantitate genomic copies of the human RNase P gene, a single-copy gene encoding the RNA moiety of the RNase P enzyme. Each well contains preloaded reaction mix (1X TaqMan Universal PCR Master Mix, RNase P primers, and FAM dye-labeled probe) and template.
TheTaqMan RNase P Instrument Verification Kit must demonstrate the ability to distinguish between 5,000 and 10,000 genomic equivalents with a 99.7% confidence level for a subsequent sample run in a single well. To ensure accurate results, we recommend that the performance of your instrument be verified every six months.
Order Info
Dry ice shipping; store plates at -15° to -25°C
Specifications
One 96-well plate for use with the 0.2 mL block. Store plate at -15 to -25°C |
|
Rnase P Instrument Verification Plate | |
Dry Ice | |
RNase P Calibration, Instrument Verification Plate |
ViiA™ 7 System, QuantStudio™ 1, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Pro, QuantStudio™ 7 Pro, QuantStudio™ 12k Flex, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex | |
Real Time PCR (qPCR) | |
1 x 96-well Plate |
For Research Use Only. Not for use in diagnostic procedures.