Learn More
Manutan Light duty mini-roller bearing alloy track, 20 roller bearings, P30, 300mm wide
Brand: Manutan A341711
This item is not returnable.
View return policy
Description
Lightweight aluminum gravity conveyor track with PVC roller bearings and mild steel bearings for parcels and packets as small as 70mm in length. Frame: 50mm x 25mm x 3mm aluminum angle, roller bearings: PVC 20mm/30mm diameter. Load capacity per metre: 25kg.
- 20mm diameter roller bearings with mild steel bearings and spindles
- Track width: 300mm
- Roller bearing pitch: 30mm
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Manutan Light duty mini-roller bearing alloy track, 20 roller bearings, P30, 300mm wide
Spot an opportunity for improvement?Share a Content Correction