Learn More
Manutan Light duty mini-roller bearing alloy track, 30 roller bearings, P35, 400mm wide
Brand:  Manutan A341695
Encompass_Preferred
Product Code. 12825578
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Lightweight aluminum gravity conveyor track with PVC roller bearings and mild steel bearings for parcels and packets as small as 70mm in length. Frame: 50mm x 25mm x 3mm aluminum angle, roller bearings: PVC 20mm/30mm diameter. Load capacity per metre: 25kg.
  • 30mm diameter roller bearings with mild steel bearings and spindles
  • Track width: 400mm
  • Roller bearing pitch: 35mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Manutan Light duty mini-roller bearing alloy track, 30 roller bearings, P35, 400mm wide