Learn More
Cole-Parmer™ Lightnin S-Series Sanitary Mixer
Lightnin S-Series Sanitary Mixers provide a truly global-designed, cost-effective mixer that meets all your cleanroom, food processing, or pharmaceutical specifications. CE area classification is certified for 'Safe Areas.'
Brand:  Cole-Parmer™ 50323-62
 View more versions of this product
Product Code. 15487473
  • 6105.00 € / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Lightnin, the industry leader in mixers, quality you can count on!
  • All stainless-steel construction for food, pharmaceutical, and cleanroom applications
  • The global, 304 SS motor comes standard with an IP55 washdown rating and is CE compliant.
Specifications
Specifications
1450 rpm
1/2 hp
316 Stainless Steel
TEFC
50 Hz
304 SS, 20-Degree Offset Clamp Mount; 316L SS Mixer Shaft; and 316L SS Impeller(s).
MS1P111041219
Sanitary Mixer