Learn More
Gilac™ Lightweight Nesting Pallet
Lightweight Pallet is nesting and therefore space-saving when stored empty.
Brand:  Gilac™ G052202
 View more versions of this product
Product Code. 17958601
  • 67.05 € / Each
Explore more special offers
Description
Description
  • Lightweight nesting plastic pallet.
  • Nesting and therefore space-saving when stored empty.
  • Storage prohibited on racks: the pads of the pallets are incompatible with the rails of the racks.
Specifications
Specifications
Aero Tote Pallet
800 x 1200 x 155 mm
1500 kg
155 mm
800 mm
1200 mm
Black
3306.93 lb.
6.1 in.
31.5 in.
47.2 in.