missing translation for 'onlineSavingsMsg'
Learn More

Linezolid, 98%

Product Code. 15502393 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15502393

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Linezolid, Zyvox, CAS # 165800-03-3 is a synthetic oxazolidinone antibiotic. | CAS: 165800-03-3 | C16H20FN3O4 | 337.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Linezolid is an organoflourine compound and an acetamide

• Linezolid acts by inhibiting initiation complex formation by binding to the 50S ribosomal subunit

Applications

• Linezolid is effective against staphylococcal and enterococcal microbes

• It is most effective against gram-positive organisms, particularly methicillin, vancomycin and penicillin resistant ones

• Suitable for research on antibiotic resistance, cross-reactivity and drug mechanisms

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 165800-03-3
Molecular Formula C16H20FN3O4
Molecular Weight (g/mol) 337.35
InChI Key TYZROVQLWOKYKF-ZDUSSCGKSA-N
Synonym linezolid, zyvox, zyvoxid, s-n-3-3-fluoro-4-morpholinophenyl-2-oxooxazolidin-5-yl methyl acetamide, zyvoxam, linezolide, linezlid, zyvoxa, linezolid usan:inn, unii-isq9i6j12j
PubChem CID 441401
ChEBI CHEBI:63607
IUPAC Name N-[[(5S)-3-(3-fluoro-4-morpholin-4-ylphenyl)-2-oxo-1,3-oxazolidin-5-yl]methyl]acetamide
SMILES CC(=O)NCC1CN(C(=O)O1)C2=CC(=C(C=C2)N3CCOCC3)F

Specifications

Melting Point 181.0°C to 183.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 250 mg
Solubility Information Solubility in water: insoluble.
Formula Weight 337.35
Percent Purity 98%
Chemical Name or Material Linezolid
compliance-icons
Product Identifier
  • Linezolid
Signal Word
  • Danger
Hazard Category
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.