missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Linolenic acid, 99%

Product Code. 10787192
Click to view available options
Quantity:
500 mg
1 g
5 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 463-40-1
Molecular Formula C18H30O2
Molecular Weight (g/mol) 278.44
MDL Number MFCD00065720
InChI Key DTOSIQBPPRVQHS-PDBXOOCHSA-N
Synonym linolenic acid, alpha-linolenic acid, linolenate, 9z,12z,15z-octadeca-9,12,15-trienoic acid, a-linolenic acid, cis,cis,cis-9,12,15-octadecatrienoic acid, all-cis-9,12,15-octadecatrienoic acid, alpha-linolenate, 9-cis,12-cis,15-cis-octadecatrienoic acid, z,z,z-9,12,15-octadecatrienoic acid
PubChem CID 5280934
ChEBI CHEBI:27432
IUPAC Name (9Z,12Z,15Z)-octadeca-9,12,15-trienoic acid
SMILES CCC=CCC=CCC=CCCCCCCCC(=O)O

Specifications

Melting Point -11.0°C
Color Colorless to Yellow
Density 0.9100g/mL
Boiling Point 230.0°C to 232.0°C (1.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3(CH2CH=CH)3(CH2)7CO2H
Refractive Index 1.48
Quantity 5 g
Beilstein 02, IV, 1781
Merck Index 15, 5561
Specific Gravity 0.91
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 278.44
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Linolenic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Linolenic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : linolenic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.