missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Advanced DMEM/F-12
GREENER_CHOICE

Advanced DMEM/F-12 (Dulbecco's Modified Eagle Medium/Ham's F-12) is a widely used basal medium that allows the culture of mammalian cells with reduced Fetal Bovine Serum (FBS) supplementation.

Brand:  Gibco™ 12634028

299.25 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.



 View more versions of this product

Product Code. 11550446

  • 331.00 € / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Requires : Supplementation
This item is not returnable. View return policy

Description

Description

Advanced DMEM/F-12 (Dulbecco's Modified Eagle Medium/Ham's F-12) is a widely used basal medium that allows the culture of mammalian cells with reduced Fetal Bovine Serum (FBS) supplementation. Compared to classic DMEM/F-12, serum supplementation can be reduced by 50–90% with no change in growth rate or morphology. Cells successfully cultured in Advanced DMEM/F-12, with no adaptation, include MRC-5, SP2, Vero, WI-38 and Jurkat.

This Advanced DMEM/F-12 is manufactured as follows:
With: Glucose, Non-essential Amino Acids, Sodium Pyruvate, Phenol Red
Without: L-glutamine, HEPES

The complete formulation is available.

Using Advanced DMEM/F-12
Advanced DMEM/F-12 is unique from other media due to addition of the following ingredients to allow for serum reduction: ethanolamine, glutathione, ascorbic acid, insulin, transferrin, AlbuMAX™ II lipid-rich bovine serum albumin for cell culture, and the trace elements sodium selenite, ammonium metavanadate, cupric sulfate, and manganous chloride. Advanced DMEM/F-12 requires supplementation with 1–5% Fetal Bovine Serum and 4 mM L-glutamine or GlutaMAX™ supplement. Many cell lines do not require adaptation to this media. The FBS concentration must be optimized for each cell line to obtain maximum serum reduction. Advanced Advanced DMEM/F-12 uses a sodium bicarbonate buffer system (3.7 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Animal Origin
Liquid
Reduced Serum
Sterile-filtered
No Glutamine, No HEPES
Gibco™
Room Temperature
MRC-5, SP2, Vero, WI-38 and Jurkat
1 X
DMEM (Dulbecco's Modified Eagle Medium)/F-12
Sterile-filtered
High Glucose, Phenol Red, Sodium Pyruvate, Non Essential Amino Acids (NEAA)
cGMP-compliant under the ISO 13485 standard
10 x 500 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Methyl benzoate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains : Methyl benzoate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.