Learn More
Gibco™ Essential 8™ Flex Medium Kit
Essential 8™ Flex is a xeno-free and feeder-free medium that supports the culture of pluripotent stem cells (PSCs) without the need for weekend feeding. Gibco™ Essential 8™ Flex Medium Kit is provided in a convenient two-component kit (500mL basal medium and 10mL supplement).
Brand:  Gibco™ A2858501
345.72 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Product Code. 15374799
  • 402.00 € / 1 set
Explore more special offers
Description
Description
  • Provides all of the benefits of Essential 8 Medium such as minimized variability of components, reliable and robust cultures with a xeno-free, cGMP produced media
  • Convenient two-component kit (500mL basal medium and 10mL supplement), and when used with vitronectin (VTN-N), provides defined system for feeder-free culture of human PSCs
  • Flexibility is an added benefit
Order Info
Approved for shipment at Room Temperature or on Wet Ice
TRUSTED_SUSTAINABILITY
Specifications
Specifications
• 500 mL Basal Medium (store at 2–8°C and protect from light)
• 10 mL Supplement (store at -5 to -20°C and protect from light)
Xeno-free
Stem Cell Media
cGMP for medical devices, 21 CFR Part 820 and ISO 13485
1 Kit
Embryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs), Human Stem Cells
Feeder-free Stem Cell Culture (Human, iPS - Induced Pluripotent Stem, Embryonic)
Phenol Red
Essential 8™
Supplement - dry ice, Basal - ambient