missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sampling Systems™ Liquid Master
Available in a range of lengths and diameters to give a range of sample volumes. Sampling Systems™ Liquid Master is ideal for taking samples from low and medium viscosity liquids.
Brand: Sampling Systems™ 5035P-2000
This item is not returnable.
View return policy
Description
- Product Contact Material Available in: 316L Stainless Steel, Food Grade PTFE/FEP, Polypropylene
- Easily stripped down for cleaning
- Single Point and Cross Sectional Sampling
Specifications
Liquid Sampler | |
420 mL | |
25 mm |
Polypropylene | |
Sampling Liquids | |
2000 mm |