missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Liquid Scintillation Vials, PET

Provides the clarity of glass with the safety of plastic.

Brand:  DWK Life Sciences 986754

247.16 EUR valid until 2024-11-29
Use promo code "21718" to get your promotional price.


 View more versions of this product

Product Code. 12457361

  • 280.00 € / Pack of 500
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Provides the clarity of glass with the safety of plastic.

  • Vials offer low permeability to solvents and minimal background counts
  • Clarity of glass with the safety of plastic
  • Vials can be safely incinerated
  • No harmful gas is generated
  • Packaged in 5 utility trays with each partition tray holding 100 vials
  • Utility trays serve as a way to store your samples
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Linerless
27 mm
20 mL
Polyethylene
22-400
Clear
PET
500
61 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers