missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Graduation for Wide-Necked Jerry Cans 10l

This Graduation for Wide-Necked Jerry Cans can be used on Buerkle™ article number 0431-0010 and 0431-1010

Brand:  Buerkle™ 0431-9011

7.84 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.


Product Code. 10419860

  • 9.45 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For wide-necked jerrycans
  • Self-adhesive strips with blue graduation marks
Specifications

Specifications

Peel and Stick
Graduation Scale
10 L Wide-Necked Jerrycans
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.