Learn More
Lithium, 99+%, granular, dry, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 180.0°C |
Boiling Point | 1340.0°C |
Density | 0.5300g/mL |
Assay Percent Range | 99+% |
Quantity | 25 g |
Fieser | 01,570; 03,174; 04,286; 05,376; 10,234; 11,286; 13,157; 15,184; 17,161 |
Merck Index | 15, 5576 |
Solubility Information | Solubility in water: reacts. Other solubilities: forming lioh and hydrogen, soluble in liquid ammonia and acids, decomposes in alcohol |
Formula Weight | 6.94 |
Percent Purity | ≥99% |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0392076 | Certificate of Analysis | 31/01/2025 | 211440250,211442500 |
A0461698 | Certificate of Analysis | 23/05/2024 | 211440250,211442500 |
A0462502 | Certificate of Analysis | 03/05/2024 | 211440250,211442500 |
A0442354 | Certificate of Analysis | 22/11/2023 | 211440250,211442500 |
A0450588 | Certificate of Analysis | 02/05/2023 | 211440250,211442500 |
Safety and Handling


- Lithium
- Danger
- Substance which\, in contact with water\, emits flammable gases Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
- P223-Do not allow contact with water.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.