missing translation for 'onlineSavingsMsg'
Learn More

Lithium acetate dihydrate, 98%, for analysis

Product Code. 15137442 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 15137442

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6108-17-4 | C2H3LiO2·2H2O | 102.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6108-17-4
Molecular Formula C2H3LiO2·2H2O
Molecular Weight (g/mol) 102.02
InChI Key IAQLJCYTGRMXMA-UHFFFAOYSA-M
Synonym lithium acetate dihydrate, lithiumacetatedihydrate, lithium acetate dihydrate, reagent grade, lioac-2h2o, lioac.2h2o, ksc498a1d, c2h3o2.li.2h2o, lithotab acetate ion dihydrate, acetic acid lithium salt dihydrate
PubChem CID 23666338
IUPAC Name lithium;acetate;dihydrate
SMILES [Li+].CC(=O)[O-].O.O

Specifications

Melting Point 56.0°C to 58.0°C
Quantity 1 kg
Assay Percent Range 98%
Linear Formula CH3CO2Li·2H2O
Merck Index 15,5577
Solubility Information Solubility in water: 290g/L (20°C). Other solubilities: very soluble in hot water, 215g/L ethanol
Formula Weight 102.02
Percent Purity 98%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Lithium acetate dihydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.