missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Aluminum Di-n-butylamide, 0.16M soln. in 1,2-Dimethoxyethane, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
Description
Lithium aluminum di-n-butylamide is used as a reagent for the determination of hydroxyl groups in silanols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Lithium aluminum di-n-butylamide is used as a reagent for the determination of hydroxyl groups in silanols.
Notes
Air, heat and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, air, heat and water.
Lithium aluminum di-n-butylamide is used as a reagent for the determination of hydroxyl groups in silanols.
Notes
Air, heat and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, air, heat and water.
Specifications
Specifications
Melting Point | -54°C |
Density | 0.86 |
Linear Formula | LiAl[N(CH2CH2CH2CH3)2]4 |
Quantity | 500 mL |
UN Number | UN1993 |
Sensitivity | Air and moisture sensitive |
Formula Weight | 546.87 |
Chemical Name or Material | Lithium Aluminum Di-n-butylamide |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction