missing translation for 'onlineSavingsMsg'
Learn More

Lithium azide, pure, 20% solution in water, Thermo Scientific Chemicals

Product Code. 10123921
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, For Azidation/Diazo transfer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19597-69-4
Molecular Formula LiN3
MDL Number MFCD00041956

Specifications

Color Colorless
Linear Formula LiN3
Density 1.0880g/mL
Formula Weight 48.96
Percent Purity 17 to 23%
Grade Pure
Packaging Glass bottle
Quantity 100 g
Physical Form Liquid
Chemical Name or Material Lithium azide, 20% Solution in Water
compliance-icons
Product Identifier
  • Lithium azide, 20% solution in water
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
Hazard Statement
  • H300-Fatal if swallowed.
  • H311-Toxic in contact with skin.
  • H330-Fatal if inhaled.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P233-Keep container tightly closed.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'