Learn More
Lithium bis(trifluoromethylsulfonyl)imide, 98+%, Thermo Scientific Chemicals
71.05 € - 234.17 €
Chemical Identifiers
CAS | 90076-65-6 |
---|---|
Molecular Formula | C2F6LiNO4S2 |
Molecular Weight (g/mol) | 287.075 |
MDL Number | MFCD00210017 |
InChI Key | QSZMZKBZAYQGRS-UHFFFAOYSA-N |
Synonym | lithium bis trifluoromethanesulfonyl imide, lithium bis trifluoromethyl sulfonyl amide, lithium bis trifluoromethanesulphonyl imide, litfsi, lithium bis trifluoromethylsulfonyl azanide, lithium bis trifluoromethylsulfonyl imide, bis trifluoromethane sulfonimide lithium salt, bistrifluoromethanesulfonimide lithium salt, lithium bis trifluoromethanesulfonyl amide, lithium triflimideShow More |
PubChem CID | 3816071 |
IUPAC Name | lithium;bis(trifluoromethylsulfonyl)azanide |
SMILES | [Li+].C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11372379
|
Thermo Scientific Alfa Aesar
H27307.09 |
10 g |
71.05 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11382379
|
Thermo Scientific Alfa Aesar
H27307.18 |
50 g |
234.17 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lithium bis(trifluoromethylsulfonyl)imide is used in the preparation of chiral imidazolium salt through an anion metathesis of the corresponding triflate organic electrolyte-based lithium batteries. It finds application in the preparation of rare-earth Lewis acid catalysts. It is also useful in primary and secondary lithium cells using organic liquid electrolytes and polymer batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsLithium bis(trifluoromethylsulfonyl)imide is used in the preparation of chiral imidazolium salt through an anion metathesis of the corresponding triflate organic electrolyte-based lithium batteries. It finds application in the preparation of rare-earth Lewis acid catalysts. It is also useful in primary and secondary lithium cells using organic liquid electrolytes and polymer batteries.
Solubility
Soluble in water.
Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Chemical Identifiers
90076-65-6 | |
287.075 | |
QSZMZKBZAYQGRS-UHFFFAOYSA-N | |
3816071 | |
[Li+].C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F |
C2F6LiNO4S2 | |
MFCD00210017 | |
lithium bis trifluoromethanesulfonyl imide, lithium bis trifluoromethyl sulfonyl amide, lithium bis trifluoromethanesulphonyl imide, litfsi, lithium bis trifluoromethylsulfonyl azanide, lithium bis trifluoromethylsulfonyl imide, bis trifluoromethane sulfonimide lithium salt, bistrifluoromethanesulfonimide lithium salt, lithium bis trifluoromethanesulfonyl amide, lithium triflimideShow More | |
lithium;bis(trifluoromethylsulfonyl)azanide |
Specifications
90076-65-6 | |
C2F6LiNO4S2 | |
MFCD00210017 | |
Moisture sensitive | |
Soluble in water. | |
[Li+].C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F | |
287.075 | |
287.08 | |
Lithium bis(trifluoromethylsulfonyl)imide |
234°C to 238°C | |
(CF3SO2)2NLi | |
UN2923 | |
lithium bis trifluoromethanesulfonyl imide, lithium bis trifluoromethyl sulfonyl amide, lithium bis trifluoromethanesulphonyl imide, litfsi, lithium bis trifluoromethylsulfonyl azanide, lithium bis trifluoromethylsulfonyl imide, bis trifluoromethane sulfonimide lithium salt, bistrifluoromethanesulfonimide lithium salt, lithium bis trifluoromethanesulfonyl amide, lithium triflimide | |
QSZMZKBZAYQGRS-UHFFFAOYSA-N | |
lithium;bis(trifluoromethylsulfonyl)azanide | |
3816071 | |
≥98% |
Safety and Handling
GHS H Statement
H301-H311-H314-H373
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301+H311-H314-H373
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II
missing translation for 'einecsNumber' : 415-300-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.