Learn More
Lithium bis(trimethylsilyl)amide, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals
56.08 € - 249.24 €
Specifications
Chemical Name or Material | Lithium bis(trimethylsilyl)amide |
---|---|
CAS | 4039-32-1, 109-99-9 |
Density | 0.9000g/mL |
Boiling Point | 65.0°C |
Flash Point | −21°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15462132
|
Thermo Scientific Acros
209551000 |
100 mL |
56.08 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15487631
|
Thermo Scientific Acros
209558000 |
800 mL |
249.24 €
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lithium bis(trimethylsilyl)amide | |
0.9000g/mL | |
−21°C | |
((CH3)3Si)2NLi | |
0.9 | |
Solubility in water: reacts. | |
[Li+].C[Si](C)(C)[N-][Si](C)(C)C | |
167.33 | |
167.33 |
4039-32-1, 109-99-9 | |
65.0°C | |
C6H18LiNSi2 | |
04,296; 05,393; 07,197; 12,280; 13,165; 14,194 | |
lithium bis trimethylsilyl amide, lithium hexamethyldisilazide, lihmds, lhmds, hexamethyldisilazane lithium salt, unii-rc4n1i108m, lithiumbis trimethylsilyl amide, lithium bis trimethylsilyl azanide, lithium hexamethyldisilazane, lithium bis-trimethylsilyl amide | |
YNESATAKKCNGOF-UHFFFAOYSA-N | |
lithium;bis(trimethylsilyl)azanide | |
2733832 |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Suspected of causing cancer.
May form explosive peroxides.
Reacts violently with water.<br/
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 223-725-6
- Lithium bis(trimethylsilyl)amide, 1M solution in THF
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.