missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium borohydride, 4M (10 wt.%) solution in THF, AcroSeal™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Specifications
Specifications
| Chemical Name or Material | Lithium Borohydride |
| CAS | 16949-15-8, 109-99-9 |
| Quantity | 100 mL |
| Molecular Formula | BH4Li |
| Linear Formula | LiBH4 |
| MDL Number | MFCD00011088 |
| Flash Point | −21°C |
| Merck Index | 15,5581 |
| Synonym | borate 1-, tetrahydro-, lithium, unii-8l87x4s4kp, boryllithium, lithium, boryl, lithium hydroborate, lithotab borohydride, bh4li, lithium 1+ ion boranuide, lithium borohydride 2.0 m in thf, borate 1-, tetrahydro-, lithium 1:1 |
| Solubility Information | Solubility in water: decomposes |
| Show More |
Safety and Handling
Product Identifier
- Lithium borohydride, 4M (10 wt.%) solution in THF
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu