missing translation for 'onlineSavingsMsg'
Learn More
Lithium bromide, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals Product Code.: 10741091

Lithium bromide, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals

Product Code. 10741091
100 g
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
This item is not returnable. View return policy

Product Code. 10741091

Brand: Thermo Scientific Acros 199871000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7550-35-8
Molecular Formula BrLi
Molecular Weight (g/mol) 86.84
MDL Number MFCD00011077
InChI Key AMXOYNBUYSYVKV-UHFFFAOYSA-M
Synonym lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide
PubChem CID 82050
ChEBI CHEBI:63042
IUPAC Name lithium;bromide
SMILES [Li+].[Br-]

Specifications

Boiling Point 1265.0°C
Melting Point 550.0°C
Color Gray to Black
pH 6 to 12 (4% soln. at 20°C)
Physical Form Powder
Quantity 100 g
Assay Percent Range 99% min. (Argentometry)
Linear Formula LiBr
Fieser 01,604; 02,245; 04,297
Merck Index 15,5582
Solubility Information Solubility in water: 1.49 kg/l (20°C). Other solubilities: slightly soluble in alcohol, ether, methanol,, acetone and glycol, slightly soluble in pyridine, soluble in amyl alcohol
Formula Weight 86.84
Percent Purity ≥99%
Grade Analytical
Loss on Drying 0.5% max. (400°C)
Packaging Plastic bottle
Specific Gravity 3.46
Density 3.4600g/mL
Chemical Name or Material Lithium bromide, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0452667 Certificate TypeCertificate of Analysis Date03/05/2023 Catalog Number199871000,199875000,199870100,199870025
Lot NumberA0402113 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number199871000,199875000,199870100,199870025
Lot NumberA0397714 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number199871000,199875000,199870100,199870025
Lot NumberA0394845 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number199871000,199875000,199870100,199870025
Lot NumberA0403586 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number199871000,199875000,199870100,199870025
compliance-icons
Product Identifier
  • Lithium bromide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lithium bromide, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.