missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Bromide, Honeywell™
Reagent Grade,≥99%
73.07 € - 410.44 €
Chemical Identifiers
CAS | 7550-35-8 |
---|---|
Molecular Formula | BrLi |
Molecular Weight (g/mol) | 86.844 |
MDL Number | MFCD00011077 |
InChI Key | AMXOYNBUYSYVKV-UHFFFAOYSA-M |
Synonym | lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide |
PubChem CID | 82050 |
ChEBI | CHEBI:63042 |
IUPAC Name | lithium;bromide |
SMILES | [Li+].[Br-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15671950
|
Honeywell Research Chemicals
213225-100G |
100 g |
73.07 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15691950
|
Honeywell Research Chemicals
213225-500G |
500 g |
134.56 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15681950
|
Honeywell Research Chemicals
213225-2KG |
2 kg |
410.44 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
7550-35-8 | |
86.844 | |
AMXOYNBUYSYVKV-UHFFFAOYSA-M | |
82050 | |
lithium;bromide |
BrLi | |
MFCD00011077 | |
lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide | |
CHEBI:63042 | |
[Li+].[Br-] |
Specifications
7550-35-8 | |
LiBr | |
NONH for all modes of transport | |
AMXOYNBUYSYVKV-UHFFFAOYSA-M | |
lithium;bromide | |
82050 | |
86.85g/mol | |
ReagentPlus™ |
BrLi | |
MFCD00011077 | |
lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide | |
[Li+].[Br-] | |
86.844 | |
CHEBI:63042 | |
≥99% | |
Lithium Bromide |
Safety and Handling
P280-P305 + P351 + P338
H302-H315-H317-H319
missing translation for 'einecsNumber' : 231-439-8
missing translation for 'rtecsNumber' : OJ5755000