missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium carbonate, Puratronic™, 99.998% (metals basis excluding Ca), Ca LT 20ppm, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1 kg
Description
Stainless Steel powder is employed in pharmaceuticals, marine applications, architectural applications, medical implants, including pins, screws and orthopaedic implants like total hip and knee replacements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Specifications
Boiling Point | 1310°C (decomposition) |
Melting Point | 723°C |
Color | White |
Physical Form | Powder |
Quantity | 250 g |
Assay Percent Range | 99.998% (Metals basis excluding Ca) |
Solubility Information | Soluble in water and acid. Insoluble in acetone,ammonia and alcohol. |
IUPAC Name | dilithium;carbonate |
Formula Weight | 73.9 |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'