missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium chloride, 99%, extra pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 1382.0°C |
Melting Point | 605.0°C |
Quantity | 1 kg |
Assay Percent Range | 98.5% min. (Argentometry) |
Linear Formula | LiCl |
Fieser | 01,609; 02,246; 04,298; 05,677; 12,277; 16,194 |
Merck Index | 15,5584 |
Solubility Information | Solubility in water: 832g/L (20°C). Other solubilities: soluble in alcohols, ether, pyridine,, nitrobenzene and acetone |
Formula Weight | 42.38 |
Percent Purity | 99% |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0447661 | Certificate of Analysis | 31/01/2025 | 193370010,193370050 |
A0468872 | Certificate of Analysis | 31/01/2025 | 193370010,193370050 |
A0467186 | Certificate of Analysis | 25/10/2024 | 193370010,193370050 |
A0465027 | Certificate of Analysis | 01/08/2024 | 193370010,193370050 |
A0458824 | Certificate of Analysis | 05/01/2024 | 193370010,193370050 |
Safety and Handling

Product Identifier
- Lithium chloride, anhydrous
- Warning
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lithium chloride, 99%, extra pure, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction