missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Chloride, ACS Reagent, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Quantity | 100 g |
Linear Formula | LiCl |
UN Number | NONH for all modes of transport |
Formula Weight | 42.39g/mol |
Percent Purity | ≥99% |
Grade | ACS Reagent |
Chemical Name or Material | Lithium Chloride |
Safety and Handling

Product Identifier
- Lithium chloride
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Lithium chloride
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lithium Chloride, ACS Reagent, Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction