missing translation for 'onlineSavingsMsg'
Learn More

Lithium chloride monohydrate, 99.95% (metals basis), Thermo Scientific Chemicals

154.77 € - 488.43 €

Chemical Identifiers

CAS 16712-20-2
Molecular Formula ClH2LiO
Molecular Weight (g/mol) 60.405
MDL Number MFCD00011078
InChI Key VXJIMUZIBHBWBV-UHFFFAOYSA-M
Synonym lithium chloride hydrate, lithium chloride monohydrate, lithium chloride, monohydrate, lithiumchloridehydrate, hcl2lio, licl h2o, licl.h2o, acmc-1bre5, ksc182g6f, lithium chloride hydrate, puratronic
PubChem CID 23681138
IUPAC Name lithium;chloride;hydrate
SMILES [Li+].O.[Cl-]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11338017
View Documents
Thermo Scientific Alfa Aesar
022151.14
25 g
154.77 €
25g
Please to purchase this item. Need a web account? Register with us today!
11328017
View Documents
Thermo Scientific Alfa Aesar
022151.22
100 g
488.43 €
100g
Please to purchase this item. Need a web account? Register with us today!
Beschreibung

Beschreibung

Lithium chloride monohydrate is used as a precursor in the production of lithium metal and other lithium compounds. It acts as a welding agent for aluminum, salt bath for heat -treatment by low temperature and soldering techniques. It is also used as a tracer for chemical products viz. denaturation of wine. For absorbers, it is an absorption and disinfection reagent as well as desiccant for air conditioner.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spezifikation

Chemische Identifikatoren

16712-20-2
60.405
VXJIMUZIBHBWBV-UHFFFAOYSA-M
23681138
[Li+].O.[Cl-]
ClH2LiO
MFCD00011078
lithium chloride hydrate, lithium chloride monohydrate, lithium chloride, monohydrate, lithiumchloridehydrate, hcl2lio, licl h2o, licl.h2o, acmc-1bre5, ksc182g6f, lithium chloride hydrate, puratronic
lithium;chloride;hydrate

Spezifikation

>98°C-H2O
White
99.95% (Metals basis)
MFCD00011078
Soluble in water.
[Li+].O.[Cl-]
60.405
60.41 (42.39 Anhydrous)
Hygroscopic
Lithium chloride monohydrate
16712-20-2
Crystalline
ClH2LiO
lithium chloride hydrate, lithium chloride monohydrate, lithium chloride, monohydrate, lithiumchloridehydrate, hcl2lio, licl h2o, licl.h2o, acmc-1bre5, ksc182g6f, lithium chloride hydrate, puratronic
VXJIMUZIBHBWBV-UHFFFAOYSA-M
lithium;chloride;hydrate
23681138
Odorless
1.78
Videos
Sicherheit und Handhabung

Sicherheit und Handhabung

P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c

H302-H315-H319

missing translation for 'einecsNumber' : 231-212-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Produktidentifikator
  • Lithium chloride monohydrate

Signalwort
  • Warning

Gefahrenkategorie
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Gefahrenhinweis
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Ergänzende Informationen
  • MIXTURE LIST-Contains : Lithium chloride, monohydrate

Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate

RUO – Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt