missing translation for 'onlineSavingsMsg'
Learn More

Lithium cobalt(III) oxide, 97%, Thermo Scientific Chemicals

79.09 € - 851.24 €

Chemical Identifiers

CAS 12190-79-3
Molecular Formula CoLiO2
Molecular Weight (g/mol) 97.871
MDL Number MFCD00049786
InChI Key BFZPBUKRYWOWDV-UHFFFAOYSA-N
Synonym lithium colbaltite, lithium cobalt iii oxide, cobalt lithium oxide, cobaltate coo21-, lithium, cobalt lithium dioxide, coo2.li, lithium oxido oxo cobalt, lithium cobalt dioxide, lithotab cobaltoylolate
PubChem CID 23670860
IUPAC Name lithium;oxido(oxo)cobalt
SMILES [Li+].[O-][Co]=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11387657
View Documents
Thermo Scientific Alfa Aesar
014049.18
50 g
79.09 €
50g
Please to purchase this item. Need a web account? Register with us today!
11307667
View Documents
Thermo Scientific Alfa Aesar
014049.30
250 g
244.22 €
250g
Please to purchase this item. Need a web account? Register with us today!
11397657
View Documents
Thermo Scientific Alfa Aesar
014049.A1
1 kg
851.24 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Aluminum oxide finds applications in catalyst support, chemical sensors, as an absorbent for gases and water vapors, manufacturing of abrasives, refractories, ceramics, electrical insulators and resistors, paper, crucibles, as chromatographic matrix, in fluxes, light bulbs and heat-resistant fiber. Heavy quantity of Aluminum oxide is used in the production of aluminum and a component of clay. It is also used as catalyst in hydrodesulfurization and in Ziegler-Natta polymerizations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Chemical Identifiers

12190-79-3
97.871
BFZPBUKRYWOWDV-UHFFFAOYSA-N
23670860
[Li+].[O-][Co]=O
CoLiO2
MFCD00049786
lithium colbaltite, lithium cobalt iii oxide, cobalt lithium oxide, cobaltate coo21-, lithium, cobalt lithium dioxide, coo2.li, lithium oxido oxo cobalt, lithium cobalt dioxide, lithotab cobaltoylolate
lithium;oxido(oxo)cobalt

Specifications

12190-79-3
Powder
CoLiO2
lithium colbaltite, lithium cobalt iii oxide, cobalt lithium oxide, cobaltate coo21-, lithium, cobalt lithium dioxide, coo2.li, lithium oxido oxo cobalt, lithium cobalt dioxide, lithotab cobaltoylolate
BFZPBUKRYWOWDV-UHFFFAOYSA-N
lithium;oxido(oxo)cobalt
23670860
Odorless
Dark blue powder
Blue
97%
MFCD00049786
Insoluble in water.
[Li+].[O-][Co]=O
97.871
97.88
Hygroscopic
Lithium cobalt(III) oxide
Videos
Safety and Handling

Safety and Handling

P201-P202-P281-P308+P313-P501c

H350-H361f

missing translation for 'einecsNumber' : 235-362-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.