Learn More
Lithium cobalt(III) oxide, 99.5% (metals basis), Thermo Scientific Chemicals
Description
Baking powder, medicine (cathartic), component of Fehling's solution, silvering mirrors.Potassium sodium L-tartrate tetrahydrate is used in the process of silvering mirrors. It is active ingredient in the Fehiling’s solution and Nylander’s test involved in the determination of reducing sugars. It is utilized to break up emulsion in organic synthesis as well as a common precipitant in protein crystallography. As a Biuret reagent, it is used to measure the protein concentration. Furthermore, it is used as laxative and is also used in food industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Specifications
Color | Blue |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 99.5% (Metal basis) |
Solubility Information | Insoluble in water. |
Formula Weight | 97.88 |
Odor | Odorless |
Sensitivity | Hygroscopic |
Appearance | Dark blue powder |
Chemical Name or Material | Lithium cobalt(III) oxide |
RUO – Research Use Only