missing translation for 'onlineSavingsMsg'
Learn More

Lithium cobalt(III) oxide, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11329737
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Baking powder, medicine (cathartic), component of Fehling's solution, silvering mirrors.Potassium sodium L-tartrate tetrahydrate is used in the process of silvering mirrors. It is active ingredient in the Fehiling’s solution and Nylander’s test involved in the determination of reducing sugars. It is utilized to break up emulsion in organic synthesis as well as a common precipitant in protein crystallography. As a Biuret reagent, it is used to measure the protein concentration. Furthermore, it is used as laxative and is also used in food industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12190-79-3
Molecular Formula CoLiO2
Molecular Weight (g/mol) 97.871
MDL Number MFCD00049786
InChI Key BFZPBUKRYWOWDV-UHFFFAOYSA-N
Synonym lithium colbaltite, lithium cobalt iii oxide, cobalt lithium oxide, cobaltate coo21-, lithium, cobalt lithium dioxide, coo2.li, lithium oxido oxo cobalt, lithium cobalt dioxide, lithotab cobaltoylolate
PubChem CID 23670860
IUPAC Name lithium;oxido(oxo)cobalt
SMILES [Li+].[O-][Co]=O

Specifications

Color Blue
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.5% (Metal basis)
Solubility Information Insoluble in water.
Formula Weight 97.88
Odor Odorless
Sensitivity Hygroscopic
Appearance Dark blue powder
Chemical Name or Material Lithium cobalt(III) oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'