missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Dodecyl Sulfate (Powder or Flakes/Molecular Biology), Fisher BioReagents
Click to view available options
Quantity:
25 g
Packaging:
Glass Bottle
Description
Lithium Dodecyl Sulfate (LDS) is a detergent used for dissociating proteins.
Specifications
Specifications
Absorbance | 0.01 max. (3% solution) at 405nm, 0.05 max. (3% solution) at 280nm, 0.100 AU max. at 260nm, 0.3 max. (3% solution) at 230nm |
Color | White |
Physical Form | Solid |
Quantity | 25 g |
Identification | Pass Test |
Packaging | Glass Bottle |
Assay Percent Range | ≥99 % |
Linear Formula | CH3(CH2)11OSO3Li |
Formula Weight | 273.18 |
Percent Purity | ≥99% (≥98% C12-OH) |
Show More |
Safety and Handling
Product Identifier
- Lithium dodecyl sulfate
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H228-Flammable solid.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction