missing translation for 'onlineSavingsMsg'
Learn More

Lithium foil, 0.75mm (0.03in) thick x 19mm (0.75in) wide, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 15424745
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7439-93-2
Assay Percent Range 99.9% (metals basis)
Molecular Formula Li
MDL Number MFCD00134051
Quantity 25 g
Synonym litio, litium, lithium, metallic, lithium, elemental, monohydride, unii-9fn79x2m3f, hsdb 647, hydride lih, hydrure de french, hsdb 549
InChI Key WHXSMMKQMYFTQS-UHFFFAOYSA-N
SMILES [Li]
IUPAC Name lithium
Molecular Weight (g/mol) 6.94
PubChem CID 3028194
ChEBI CHEBI:30145
Chemical Name or Material Lithium foil
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Lithium foil
Signal Word
  • Danger
Hazard Category
  • Substance which\, in contact with water\, emits flammable gases Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P223-Do not allow contact with water.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'