missing translation for 'onlineSavingsMsg'
Learn More
Lithium hydroxide, 98%, pure, anhydrous, Thermo Scientific Chemicals Product Code.: 10728213

Lithium hydroxide, 98%, pure, anhydrous, Thermo Scientific Chemicals

Product Code. 10728213
100 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10728213

Brand: Thermo Scientific Acros 379181000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-65-2
Molecular Formula HLiO
Molecular Weight (g/mol) 23.95
MDL Number MFCD00011095
InChI Key WMFOQBRAJBCJND-UHFFFAOYSA-M
Synonym lithium hydroxide, lithium hydrate, lioh, lithium hydroxide anhydrous, lithium hydroxide li oh, lithiumhydroxid, lithiumhydroxide, lithium hydoxide, unii-903yl31jas, lithium hydroxide, anhydrous
PubChem CID 3939
ChEBI CHEBI:33979
SMILES [Li+].[OH-]

Specifications

Melting Point 462.0°C
Color White to Yellow
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 98%
Merck Index 15,559
Solubility Information Solubility in water: 113g/L (20°C). Other solubilities: slightly soluble in alcohol
IUPAC Name lithium;hydroxide
Formula Weight 23.95
Percent Purity 98%
Grade Pure
Packaging Plastic bottle
Specific Gravity 1.43
Density 1.4300g/mL
Chemical Name or Material Lithium hydroxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Lithium hydroxide anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lithium hydroxide, 98%, pure, anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.