missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Metaborate (100%), Fusion Flux Pure Crystal For Sample Preparation, SPEX CertiPrep™
Click to view available options
Quantity:
1 kg
Packaging:
Triple-leached LDPE bottle
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
- Pure Grade Fusion Flux
- 1 kg
- Lithium Metaborate (100%)
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Compliance
GHS/OSHA Complaint
Specifications
Specifications
CAS | 13453-69-5 |
Packaging | Triple-leached LDPE bottle |
Quantity | 1 kg |
Certificate of Analysis | With Certificate of Analysis. |
Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Molecular Formula | B4H14Li2O7 |
InChI Key | DPYKRXYVXDYLEY-UHFFFAOYSA-N |
SMILES | [Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O |
IUPAC Name | tetraboron heptahydrate dilithium |
Show More |
Safety and Handling
Product Identifier
- BORATE FUSION FLUX
- Warning
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Product intended for laboratory use only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'