missing translation for 'onlineSavingsMsg'
Learn More

Lithium methoxide, 98+%, Thermo Scientific Chemicals

78.49 € - 982.89 €

Chemical Identifiers

CAS 865-34-9
Molecular Formula CH3LiO
Molecular Weight (g/mol) 37.97
MDL Number MFCD00036357
InChI Key JILPJDVXYVTZDQ-UHFFFAOYSA-N
Synonym lithium methoxide, lithium methanolate, methanol, lithium salt, lithium methylate, liome, methanol, lithium salt 1:1, liom, lithium methylat, lithotab methoxide, lioch3
PubChem CID 4177230
SMILES [Li+].C[O-]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11309618
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044129.14
25 g
78.49 €
25g
Please to purchase this item. Need a web account? Register with us today!
11389608
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044129.22
100 g
239.19 €
100g
Please to purchase this item. Need a web account? Register with us today!
11399608
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044129.36
500 g
982.89 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Lithium methoxide is used in sol-gel synthesis of lithium borophosphates. It is also useful for deprotonation reactions in organic synthesis. Further, it serves as a mild base in organic synthesis especially in transesterifications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

865-34-9
37.97
JILPJDVXYVTZDQ-UHFFFAOYSA-N
4177230
CH3LiO
MFCD00036357
lithium methoxide, lithium methanolate, methanol, lithium salt, lithium methylate, liome, methanol, lithium salt 1:1, liom, lithium methylat, lithotab methoxide, lioch3
[Li+].C[O-]

Specifications

865-34-9
White
LiOCH3
UN3206
Air and moisture sensitive
Soluble in methanol.
[Li+].C[O-]
37.97
37.98
Powder
>500°C (decomposition)
CH3LiO
MFCD00036357
3552389
lithium methoxide, lithium methanolate, methanol, lithium salt, lithium methylate, liome, methanol, lithium salt 1:1, liom, lithium methylat, lithotab methoxide, lioch3
JILPJDVXYVTZDQ-UHFFFAOYSA-N
lithium(1+) methanolate
4177230
≥98%
Lithium methoxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H251-H314
Self-heating: may catch fire.
Causes severe skin burns and eye damage.

P235+P410-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-X

missing translation for 'dotInformation' : Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: ALKALI METAL ALCOHOLATES, SELF-HEATING, CORROSIVE, N.O.S.

missing translation for 'einecsNumber' : 212-737-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only