missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Nitrate Anhydrous, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
100 g
500 g
5 kg
Specifications
Specifications
Boiling Point | 600°C |
Melting Point | 251°C |
Color | White |
Physical Form | Solid |
Quantity | 5 kg |
IUPAC Name | lithium(1+) nitrate |
Formula Weight | 68.95g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'