missing translation for 'onlineSavingsMsg'
Learn More
Lithium Nitrate, Reagent Grade, Honeywell™ Product Code.: 16695763

Lithium Nitrate, Reagent Grade, Honeywell™

Product Code. 16695763
25 kg
Click to view available options
Quantity:
25 kg
50 kg
This item is not returnable. View return policy

Product Code. 16695763

Brand: Honeywell Research Chemicals 22798625KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7790-69-4
Molecular Formula LiNO3
Molecular Weight (g/mol) 68.94
MDL Number MFCD00011094
InChI Key IIPYXGDZVMZOAP-UHFFFAOYSA-N
IUPAC Name lithium(1+) nitrate
SMILES [Li+].[O-][N+]([O-])=O

Specifications

Boiling Point >450°C (Decomposes below the boiling point)
Melting Point 0.25°C 3 to 264°C
Color Colorless
CAS Max % ≤ 100%
pH 7 to 9
Quantity 25 kg
Density 2.38 g/cm3
Chemical Name or Material Lithium nitrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lithium Nitrate, Reagent Grade, Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.