Learn More
Lithium perchlorate trihydrate, 99%, extra pure, Thermo Scientific Chemicals
28.44 € - 299.49 €
Chemical Identifiers
CAS | 13453-78-6 |
---|---|
Molecular Formula | ClLiO4·3H2O |
Molecular Weight (g/mol) | 160.44 |
MDL Number | MFCD00149765 |
InChI Key | JAWYRNYHJJDXHX-UHFFFAOYSA-M |
Synonym | lithium perchlorate trihydrate, perchloric acid, lithium salt, trihydrate, unii-jyk745743v, acmc-1c1zx, lithium perchlorate hydrate, lithiumperchlorate trihydrate, ksc491a4p, lithium pechlorate trihydrate, lithiumperchloratetrihydrate, lithotab trihydrate perchlorate ion |
PubChem CID | 23708903 |
IUPAC Name | lithium;perchlorate;trihydrate |
SMILES | [Li+].O.O.O.[O-]Cl(=O)(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10582981
|
Thermo Scientific Acros
194720250 |
25 g | Plastic bottle |
28.44 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10032530
|
Thermo Scientific Acros
194722500 |
250 g | Plastic bottle |
135.68 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10637694
|
Thermo Scientific Acros
194720010 |
1 kg | Plastic bottle |
299.49 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
13453-78-6 | |
160.44 | |
JAWYRNYHJJDXHX-UHFFFAOYSA-M | |
23708903 | |
[Li+].O.O.O.[O-]Cl(=O)(=O)=O |
ClLiO4·3H2O | |
MFCD00149765 | |
lithium perchlorate trihydrate, perchloric acid, lithium salt, trihydrate, unii-jyk745743v, acmc-1c1zx, lithium perchlorate hydrate, lithiumperchlorate trihydrate, ksc491a4p, lithium pechlorate trihydrate, lithiumperchloratetrihydrate, lithotab trihydrate perchlorate ion | |
lithium;perchlorate;trihydrate |
Specifications
95°C | |
White | |
Crystals and/or Chunks | |
ClLiO4·3H2O | |
MFCD00149765 | |
lithium perchlorate trihydrate, perchloric acid, lithium salt, trihydrate, unii-jyk745743v, acmc-1c1zx, lithium perchlorate hydrate, lithiumperchlorate trihydrate, ksc491a4p, lithium pechlorate trihydrate, lithiumperchloratetrihydrate, lithotab trihydrate perchlorate ion | |
JAWYRNYHJJDXHX-UHFFFAOYSA-M | |
lithium;perchlorate;trihydrate | |
23708903 | |
99% | |
Lithium perchlorate trihydrate, p.a. |
13453-78-6 | |
6.0 to 7.5 (5% soln. at 20°C) | |
98.5% min. (Argentometry) | |
LiClO4·3H2O | |
15, 5595 | |
Solubility in water: 597g/L water (25°C). Other solubilities: soluble in alcohol | |
[Li+].O.O.O.[O-]Cl(=O)(=O)=O | |
160.44 | |
160.44 | |
Extra Pure |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May intensify fire; oxidizer.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'rtecsNumber' : SC8750000
- Lithium perchlorate trihydrate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H272-May intensify fire; oxidiser.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.