Learn More
Lithium standard solution, for AAS, 1 mg/ml Li in 2% HCl, Thermo Scientific Chemicals
36.18 € - 108.54 €
Specifications
Chemical Name or Material | Lithium standard solution |
---|---|
Concentration | 995 to 1005 μg/mL of Li (1+) in 2% HCl (20°C) |
Name Note | 1 mg/ml Li in 2% HCl |
CAS | 7439-93-2, 7647-01-0, 7732-18-5 |
Packaging | Plastic bottle |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10286680
|
Thermo Scientific Acros
196081000 |
100 mL | Plastic bottle |
36.18 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10388330
|
Thermo Scientific Acros
196085000 |
500 mL | Plastic bottle |
108.54 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopySpecifications
Lithium standard solution | |
1 mg/ml Li in 2% HCl | |
Plastic bottle | |
MFCD00134051 | |
[Li] | |
6.94 | |
CHEBI:30145 | |
AAS | |
Solution |
995 to 1005 μg/mL of Li (1+) in 2% HCl (20°C) | |
7439-93-2, 7647-01-0, 7732-18-5 | |
Li | |
WHXSMMKQMYFTQS-UHFFFAOYSA-N | |
lithium | |
3028194 | |
6.94 | |
0.53 |
Safety and Handling
GHS H Statement:
May be corrosive to metals.
GHS P Statement:
Keep only in original container.
Absorb spillage to prevent material damage.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
- Lithium atomic absorption standard solution
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P234-Keep only in original container.
- P390-Absorb spillage to prevent material damage.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.