missing translation for 'onlineSavingsMsg'
Learn More

Lithium Sulfate Monohydrate, Extra Pure, SLR, Fisher Chemical

Product Code. 10355950
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Melting Point 860°C
CAS 10102-25-7
Color White
Physical Form Powder/Solid
Quantity 500 g
Molecular Formula H2Li2O5S
MDL Number MFCD00149766
Synonym lithium sulfate monohydrate, dilithium 1+ ion hydrate sulfate, dilithium sulfate hydrate, sufuric acid, dilithium slat, monohydrate, lithiumsulfatemonohydrate, dilithotab hydrate sulfate, lithium sulphate monohydrate, lithium sulfate, acs, ksc181s7n
InChI Key RKGLUDFWIKNKMX-UHFFFAOYSA-L
SMILES [Li+].[Li+].O.[O-]S([O-])(=O)=O
IUPAC Name dilithium;sulfate;hydrate
Molecular Weight (g/mol) 127.95
PubChem CID 165815
Formula Weight 127.96g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • Lithium sulfate monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'