missing translation for 'onlineSavingsMsg'
Learn More

Lithium triethylborohydride, 1.7M solution in THF, Thermo Scientific Chemicals

Product Code. 15103498
Click to view available options
Quantity:
100 mL
800 mL
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22560-16-3
Molecular Formula C6H16BLi
Molecular Weight (g/mol) 105.94
MDL Number MFCD00011703
InChI Key WCJAYABJWDIZAJ-UHFFFAOYSA-N
IUPAC Name lithium(1+) triethylboranuide
SMILES [Li+].CC[BH-](CC)CC

Specifications

Linear Formula LiB(C2H5)3H
Density 0.8900g/mL
Solubility Information Solubility in water: reacts.
Formula Weight 105.94
Specific Gravity 0.89
Quantity 100 mL
Flash Point −17°C
Chemical Name or Material Lithium triethylborohydride
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lithium triethylborohydride, 1.7M solution in THF
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Pyrophoric liquid Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H250-Catches fire spontaneously if exposed to air.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'