missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Litmus Paper Roll

Product Code. p-8658026 Shop All Fisherbrand Products
Click to view available options
Type:
Litmus Paper, Blue
Litmus Paper, Red
Unit Size:
4 units

Product Code. 15980872

Please to purchase this item. Need a web account? Register with us today!

Indicator papers without colour scale. For orientation and adjustment of pH values.

  • Impregnated indicator papers
  • Allows pH adjustment during special chemical procedures, not exact pH determination
  • No colour scale provided

Specifications

pH Range 5 to 8
Format Roll
For Use With (Application) For orientation and adjustment of pH values
Type Litmus Paper, Blue
Length (Metric) 5 m
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.