missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Litmus, Pure, Indicator Grade, Fisher Chemical
Click to view available options
Quantity:
25 g
Packaging:
Glass Jar
Specifications
Specifications
CAS | 1393-92-6 |
MDL Number | 131528 |
Packaging | Glass Jar |
Color | Blue |
Quantity | 25 g |
Physical Form | Solid |
Safety and Handling
Product Identifier
- Litmus
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction