missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ LIVE/DEAD™ Fixable Blue Dead Cell Stain Kit, for UV excitation

The LIVE/DEAD™ Fixable Blue Dead Cell Stain Kit is used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining, or prior to elimination of biohazardous materials using formaldehyde fixation.

Brand:  Molecular Probes™ L34961

 View more versions of this product

Product Code. 15582241

  • 205.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining or prior to elimination of biohazardous materials using formaldehyde fixation
  • Kit has been optimized and validated for use with a UV laser flow cytometer
  • Conveniently packaged in 40-test vials to help ensure the stability and performance of the dye over time
  • Amine reactive dyes in solution will lose their effectiveness over a short period of time, therefore it is recommended to completely use the vial once rehydrated
  • If this is not possible, aliquot the vials in small volumes and store at -80°C, avoiding freeze-thaw cycles
  • LIVE/DEAD™ Fixable Dead Cell Stains are available in a wide variety of colors to meet multi-color panel needs

Optimal brightness

  • The LIVE/DEAD™ Fixable Blue Stain was selected based on its fluorescent properties to provide a bright signal when excited with a UV laser
  • The blue-fluorescent reactive dye has an excitation maximum of ∼350 nm, making it ideal for use with an UV laser, and an emission of ∼450 nm
  • By switching viability staining from a channel off of the blue 488 nm laser to the UV laser, a channel is freed up on the flow cytometer for other reagents that are difficult to find in colors other than FITC or PE
  • By using the UV laser, there is no spectral overlap issues with other common dyes, thus eliminating the need to compensate the viability stains from other stains in the panel
  • Since live and dead cells can be discriminated using a single dye and a single channel of a flow cytometer, it is an ideal choice for multi-color experiments
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Impermeant
Blue
LIVE/DEAD™ Fixable Blue Dead Cell Stain Kit, for UV excitation
Viability Assay
Dead Cell Stain Kit
450 nm
Solid
LIVE/DEAD™
Room Temperature
Eukaryotic Cells
Contains 2 vials of LIVE/DEAD™ fixable dead cell stain and 500 μL DMSO.

Store at -20°C.
Fluorescence
Flow Cytometer
LIVE/DEAD™ Fixable Blue Dead Cell Stain
350 nm
Tube(s)
80 Assays
DMSO (Dimethylsulfoxide)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3011776 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog NumberL34961
Lot Number2765739 Certificate TypeCertificate of Analysis Date07/11/2023 Catalog NumberL34961
Lot Number2559165 Certificate TypeCertificate of Analysis Date24/01/2023 Catalog NumberL34961
Lot Number2383838 Certificate TypeCertificate of Analysis Date04/03/2022 Catalog NumberL34961
Lot Number2356793 Certificate TypeCertificate of Analysis Date26/05/2021 Catalog NumberL34961
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molecular Probes™ LIVE/DEAD™ Fixable Blue Dead Cell Stain Kit, for UV excitation > 80 assays

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.