missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LIVE/DEAD™ Fixable Orange (602) Viability Kit, for 561 nm excitation
The LIVE/DEAD Fixable viability kits allow for clear determination of live and dead cells, enabling more precise analysis with both conventional and spectral flow cytometry.
201.00 € - 528.00 €
Specifications
Detection Method | Fluorescence |
---|---|
Conjugate | LIVE/DEAD Fixable Orange (602) Cell Stain |
For Use With (Application) | Flow Cytometry |
Description | LIVE/DEAD™ Fixable Orange (602) Viability Kit, for 561 nm excitation |
Format | Tubes |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17438262
|
Invitrogen™
L34983 |
80 Assays |
201.00 €
Pack of 80 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17458262
|
Invitrogen™
L34985 |
400 Assays |
528.00 €
Pack of 400 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Fixable viability kits allow for clear determination of live and dead cells, enabling more precise analysis with both conventional and spectral flow cytometry
- Cells labeled with LIVE/DEAD Fixable viability dyes can be washed, fixed, permeabilized, and stained for intracellular antigens while maintaining distinct live and dead cell populations
- The narrow and unique emission spectra are ideal for expanding high-parameter flow cytometry experiments
- Designed to provide a bright signal with minimal overlap into adjacent channels, simplifying spectral compensation
- Since live and dead cells can be discriminated using a single flow cytometer channel, these viability dyes are an ideal choice for multi-color experiments.
- Their unique spectral fingerprints are a valuable addition to spectral flow cytometry experiments
- Dyes have been conveniently packaged in single-use vials to help ensure dye stability and performance over time
- Dissolve the dye in the included anhydrous DMSO and use immediately, or store at -20°C, avoiding freeze-thaw cycles
- The LIVE/DEAD Fixable Dead Cell Stain Sampler Kit provides one vial (40 assays) of each of eight available dead cell stains
Specifications
Fluorescence | |
Flow Cytometry | |
Tubes | |
Eukaryotic Cells | |
561 nm | |
Room Temperature | |
Flow Cytometer |
LIVE/DEAD Fixable Orange (602) Cell Stain | |
LIVE/DEAD™ Fixable Orange (602) Viability Kit, for 561 nm excitation | |
Orange | |
578/602 nm | |
DMSO (Dimethylsulfoxide) | |
LIVE/DEAD™ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title