missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LIVE/DEAD™ Fixable Red Dead Cell Stain Kit, for 488 nm excitation

The LIVE/DEAD™ Fixable Red Dead Cell Stain Kit is used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining, or prior to elimination of biohazardous materials using formaldehyde fixation.

Brand:  Invitrogen™ L34972

 View more versions of this product

Product Code. 15638900

  • 544.00 € / Pack of 400

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining or prior to elimination of biohazardous materials using formaldehyde fixation
  • Kit has been optimized and validated for use with a blue laser flow cytometer
  • Conveniently packaged in 40-test vials to help ensure the stability and performance of the dye over time
  • Amine reactive dyes in solution will lose their effectiveness over a short period of time, therefore it is recommended to completely use the vial once rehydrated
  • If this is not possible, aliquot the vials in small volumes and store at -80°C, avoiding freeze-thaw cycles
  • LIVE/DEAD™ Fixable Dead Cell Stains are available in a wide variety of colors to meet multi-color panel needs

Optimal brightness

  • The LIVE/DEAD™ Fixable Red stain was selected based on its fluorescent properties to provide a bright signal when excited with a yellow, green, or blue 488 nm laser
  • The red-fluorescent reactive dye has an excitation maximum of ∼595 nm, making it ideal for use with the yellow laser, and an emission of ∼615 nm, allowing collection in the 610 or 630 BP filter
  • LIVE/DEAD™ Fixable Red Dead Cell stain is also excited well with the blue 488 nm laser
  • Since live and dead cells can be discriminated using a single dye and a single channel of a flow cytometer, it is an ideal choice for multi-color experiments
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Impermeant
Red
LIVE/DEAD™ Fixable Red Dead Cell Stain Kit, for 488 nm excitation
Viability Assay
Stain
615 nm
Solid
LIVE/DEAD™
Room Temperature
Eukaryotic Cells
Contains 10 vials of LIVE/DEAD™ fixable dead cell stain and 500 μL DMSO.

Store at -20°C.
Fluorescence
Flow Cytometer
LIVE/DEAD™ Fixable Red Dead Cell Stain
488, 595 nm
Tube(s)
400 Assays
DMSO (Dimethylsulfoxide)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.