missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LIVE/DEAD™ Viability/Cytotoxicity Assay Kit (Green/Deep Red)

A two-color fluorescent cell viability assay that simultaneously identifies live and dead cells. Live cells are labeled with calcein dye and emit a green fluorescence, while dead cells are labeled with SYTOX Deep Red dye and emit a red fluorescence.

Brand:  Invitrogen™ L32250

187.60 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.


Product Code. 17444512

  • 268.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

LIVE/DEAD™ Viability/Cytotoxicity Assay Kit (Green/Deep Red)

  • A two-color fluorescent cell viability assay that simultaneously identifies live and dead cells
  • Live cells are labeled with calcein dye and emit a green fluorescence
  • Dead cells are labeled with SYTOX Deep Red dye and emit a red fluorescence
  • Dyes can be multiplexed with other orange/red fluorophores with compatible filter sets
  • Kit is suitable for use with fluorescence microscopes or high content analysis (HCA) fluorescence multiwall plate scanners
  • Easily adaptable for use other fluorescence detection systems
  • Assay principles are general and applicable to most eukaryotic cell types, including adherent and non-adherent cells
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Green, Deep Red
LIVE/DEAD Viability/Cytotoxicity Assay Kit (Green/Deep Red)
Viability Assay
M25
495, 660 nm
Calcein, AM (live cell indicator), SYTOX™ Deep Red Nucleic Acid Stain (dead cell indicator)
2 x 40 μL Vials kit
Viability
• 1 vial of Calcein AM (live cell indicator), 40 μL, provided as a 2000X (4 mM) solution in DMSO
• 1 vial of SYTOX Deep Red Nucleic Acid Stain (dead cell indicator), 40 μL, provided as a 2000X (500 μM) solution in DMSO

Store at ≤-20°C, upright in dry place. Protect from light.
Fluorescence
Fluorescence Microscope, High Content Analysis Instrument, Other Fluorescence Detection Systems
515, 682 nm
Kit
LIVE/DEAD
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2975922 Certificate TypeCertificate of Analysis Date03/09/2024 Catalog NumberL32250
Lot Number2856035 Certificate TypeCertificate of Analysis Date29/02/2024 Catalog NumberL32250
Lot Number2776011 Certificate TypeCertificate of Analysis Date13/10/2023 Catalog NumberL32250
Lot Number2626247 Certificate TypeCertificate of Analysis Date21/06/2023 Catalog NumberL32250
Lot Number2566207 Certificate TypeCertificate of Analysis Date25/01/2023 Catalog NumberL32250
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ LIVE/DEAD™ Viability/Cytotoxicity Assay Kit (Green/Deep Red) >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.