Learn More
Heinz Herenz LMS-Swab, Amies
Test tube with the pink screw cap containing 1 mL of Amies medium for sampling and preservation of aerobic, anaerobic, and fastidious bacteria in 48 hours.
Brand:  Heinz Herenz 1020204
 View more versions of this product
Product Code. 17927842
  • 817.00 € / Pack of 900
Explore more special offers
Description
Description
  • For transportation and preservation of microbiological samples
  • Liquid media allows for easy dilution of samples
  • Carry out differnt tests out of one sample
  • Ready-to-use device delivered in an easy-to-open peel pouch
  • Breakpoint in the shaft if the applicator allows for a safe and hygienic work-flow
  • Pre-labeled test tubes allow for correct identification
  • Produced according to GMP (Good Manufacturing Practice) and certified according to DIN EN ISO 13485 ensure the high quality
  • Sterile (SAL 10-6)
Specifications
Specifications
Transport and Collection Swab
Pink
Test tube with screw cap and liquid media
Foam stick
Sterile
Pyrogen-free and non-cytotoxic
Peel pouch
Fine